(±)-4-Chlorostyrene oxide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(4-Chlorophenyl)oxirane

H26516-06 H26516-14
AAH26516-06EA 78.91 USD
AAH26516-06 AAH26516-14
(±)-4-Chlorostyrene oxide 98%
(±)-4-Chlorostyrene oxide
Formula: C₈H₇ClO
MW: 154.6 g/mol
Boiling Pt: 230 °C
Density: 1.283
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00956179
CAS Number: 2788-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR