Thiazolidine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H26427-03
H26427-06
AAH26427-06EA
124.99
USD
AAH26427-06
AAH26427-03
Thiazolidine 97%
Thiazolidine
Formula:
C₃H₇NS Boiling Pt: 165 °C Density: 1.131 Flash Pt: 56 °C(133 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005211 CAS Number: 504-78-9 EINECS: 208-002-5 UN: 2924 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...