4-Chloro-2,6-difluorobenzonitrile 97%

Warning

H26298-03 H26298-06
AAH26298-03EA 131.15 USD
AAH26298-03 AAH26298-06
4-Chloro-2,6-difluorobenzonitrile 97%
4-Chloro-2,6-difluorobenzonitrile
Formula: C₇H₂ClF₂N
MW: 173.55 g/mol
Melting Pt: 100…103 °C
Storage Temperature: Ambient
MDL Number: MFCD06660237
CAS Number: 886500-41-0
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR