2,3-Dimethylpiperazine cis- and trans- mixture 95%
|
Danger
|
H26252-MD
H26252-03
AAH26252-MDEA
108.72
USD
AAH26252-MD
AAH26252-03
2,3-Dimethylpiperazine cis- and trans- mixture 95%
2,3-Dimethylpiperazine
Formula:
C₆H₁₄N₂ Melting Pt: 81…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD07373397 CAS Number: 84468-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...