2-Methylheptane 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Isoheptane

H26220-06 H26220-14 H26220-03
AAH26220-06EA 171.99 USD
AAH26220-06 AAH26220-14 AAH26220-03
2-Methylheptane 99%
2-Methylheptane
Formula: C₈H₁₈
MW: 114.23 g/mol
Boiling Pt: 116 °C
Melting Pt: –109 °C
Density: 0.698
Flash Pt: 4 °C(40 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009518
CAS Number: 592-27-8
EINECS: 209-747-9
UN: 1262
ADR: "3",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR