4-Bromo-3,5-difluoroaniline 99%

Warning

H26201-06 H26201-03
AAH26201-06EA 182.04 USD
AAH26201-06 AAH26201-03
4-Bromo-3,5-difluoroaniline 99%
4-Bromo-3,5-difluoroaniline
Formula: C₆H₄BrF₂N
MW: 208.01 g/mol
Melting Pt: 99…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00236593
CAS Number: 203302-95-8
EINECS: None
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR