(4-Cyanophenyl)acetic acid ethyl ester 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 2-(4-cyanophenyl)acetate
, Ethyl (4-cyanophenyl)acetate
H26078-06
H26078-03
AAH26078-03EA
113.99
USD
AAH26078-03
AAH26078-06
(4-Cyanophenyl)acetic acid ethyl ester 98%
(4-Cyanophenyl)acetic acid ethyl ester
Formula:
C₁₁H₁₁NO₂ MW: 189.21 g/mol Melting Pt: 90…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD09953133 CAS Number: 1528-41-2 UN: 3439 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...