α,α-Difluorophenylacetic acid 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
α,α-Difluorobenzeneacetic acid
H26028-MD
AAH26028-MDEA
64.79
USD
AAH26028-MD
α,α-Difluorophenylacetic acid 99%
α,α-Difluorophenylacetic acid
Formula:
C₈H₆F₂O₂ MW: 172.13 g/mol Melting Pt: 73…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00498146 CAS Number: 360-03-2 EINECS: 623-610-4 UN: 3261 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...