1,3,5-Tribenzylhexahydro-1,3,5-triazine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,3,5-Tribenzyl-1,3,5-triazinane
H25964-14
H25964-06
AAH25964-14EA
165.99
USD
AAH25964-14
AAH25964-06
1,3,5-Tribenzylhexahydro-1,3,5-triazine 98+%
1,3,5-Tribenzylhexahydro-1,3,5-triazine
Formula:
C₂₄H₂₇N₃ MW: 357.5 g/mol Boiling Pt: 100 °C/0.005 mm Melting Pt: 50…51 °C Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014599 CAS Number: 2547-66-2 EINECS: 219-831-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...