Acetoxyacetyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acetoxyacetylchloride2-Chloro-2-oxoethyl acetate

H25797-06 H25797-14
AAH25797-14EA 97.29 USD
AAH25797-14 AAH25797-06
Acetoxyacetyl chloride 97%
Acetoxyacetyl chloride
Formula: C₄H₅ClO₃
MW: 136.53 g/mol
Boiling Pt: 55 °C/12 mm
Density: 1.270
Flash Pt: 71 °C(160 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011535
CAS Number: 13831-31-7
EINECS: 237-542-4
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR