DL-Methionine ethyl ester hydrochloride 98+%

H25781-06 H25781-14
AAH25781-14EA 116.55 USD
AAH25781-14 AAH25781-06
DL-Methionine ethyl ester hydrochloride 98+%
DL-Methionine ethyl ester hydrochloride
Formula: C₇H₁₆ClNO₂S
Melting Pt: 72…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00237859
CAS Number: 6297-53-6
EINECS: 228-571-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR