1-(2,4-Dimethylphenyl)-2-urea 98%

Synonyms: 2,4-Dimethylphenylurea

B25668-06 B25668-14 B25668-14 B25668-06
AAAB25668-14EA 69.31 USD
AAAB25668-14 CAAAAB25668-06 AAAB25668-06 CAAAAB25668-14
1-(2,4-Dimethylphenyl)-2-urea 98%
1-(2,4-Dimethylphenyl)-2-urea
Formula: C₉H₁₂N₂O
Melting Pt: 203…204 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00025409
CAS Number: 2990-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR