Sarcosine hydrochloride 99%

Synonyms: 2-(Methylamino)acetic acid hydrochloride

B25536-14 B25536-22 B25536-22 B25536-14
AAAB25536-14EA 68.26 USD
CAAAAB25536-22 AAAB25536-14 AAAB25536-22 CAAAAB25536-14
Sarcosine hydrochloride 99%
Sarcosine hydrochloride

A type 1 glycine transporter inhibitor

Formula: C₃H₈ClNO₂
MW: 125.55 g/mol
Melting Pt: ca 173 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012609
CAS Number: 637-96-7
EINECS: 211-310-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR