Diethyl isopropylmalonate 98+%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: Diethyl Isopropylmalonate

B25384-18 B25384-09 B25384-18 B25384-09
AAAB25384-18EA 138 USD
CAAAAB25384-18 CAAAAB25384-09 AAAB25384-18 AAAB25384-09
Diethyl isopropylmalonate 98+%
Diethyl isopropylmalonate
Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
Boiling Pt: 102…104 °C/12 mm
Density: 0,981
Flash Pt: 92 °C(197 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040491
CAS Number: 759-36-4
EINECS: 212-068-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR