N-(4-Iodophenyl)maleamic acid 97%

Warning

B25210-03 B25210-03 B25210-06 B25210-06
AAAB25210-06EA 71.93 USD
AAAB25210-06 CAAAAB25210-06 AAAB25210-03 CAAAAB25210-03
N-(4-Iodophenyl)maleamic acid 97%
N-(4-Iodophenyl)maleamic acid
Formula: C₁₀H₈INO₃
Melting Pt: 193…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00020473
CAS Number: 17280-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR