4,5-Diphenyl-1,2,3-thiadiazole 97%

B25140-03 B25140-03 B25140-06 B25140-06
AAAB25140-06EA 174.27 USD
CAAAAB25140-06 AAAB25140-06 CAAAAB25140-03 AAAB25140-03
4,5-Diphenyl-1,2,3-thiadiazole 97%
4,5-Diphenyl-1,2,3-thiadiazole
Formula: C₁₄H₁₀N₂S
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00082687
CAS Number: 5393-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR