4-Chlorophenylurea 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(4-Chlorophenyl)urea

B25090-22 B25090-22 B25090-14 B25090-14 B25090-06 B25090-06
AAAB25090-14EA 163 USD
CAAAAB25090-06 AAAB25090-14 AAAB25090-22 CAAAAB25090-14 CAAAAB25090-22 AAAB25090-06
4-Chlorophenylurea 98%
4-Chlorophenylurea
Formula: C₇H₇ClN₂O
MW: 170.6 g/mol
Melting Pt: 204…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00014788
CAS Number: 140-38-5
EINECS: 205-412-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR