N-(2,4-Dichlorophenyl)maleamic acid 97%

Warning

B24941-14 B24941-03 B24941-06 B24941-14 B24941-03 B24941-06
AAAB24941-03EA 30.72 USD
AAAB24941-03 AAAB24941-14 CAAAAB24941-06 CAAAAB24941-14 CAAAAB24941-03 AAAB24941-06
N-(2,4-Dichlorophenyl)maleamic acid 97%
N-(2,4-Dichlorophenyl)maleamic acid
Formula: C₁₀H₇Cl₂NO₃
Melting Pt: 177…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00082905
CAS Number: 95695-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR