2-Benzylidene-1-tetralone 97%

B24699-06 B24699-06 B24699-03 B24699-03
AAAB24699-03EA 61.22 USD
CAAAAB24699-03 AAAB24699-03 CAAAAB24699-06 AAAB24699-06
2-Benzylidene-1-tetralone 97%
2-Benzylidene-1-tetralone
Formula: C₁₇H₁₄O
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00019675
CAS Number: 6261-32-1
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR