1-(Dimethylamino)-2-propanol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: DimepranolN,N-DimethylisopropanolamineN,N-Dimethyl-1-amino-2-propanol

B24234-30 B24234-0B
AAB24234-0BEA 109.99 USD
AAB24234-0B AAB24234-30
1-(Dimethylamino)-2-propanol 99%
1-(Dimethylamino)-2-propanol
Formula: C₅H₁₃NO
MW: 103.16 g/mol
Boiling Pt: 123…126 °C
Melting Pt: –85 °C
Density: 0.85
Flash Pt: 35 °C(95 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004532
CAS Number: 108-16-7
EINECS: 203-556-4
UN: 2734
ADR: "8",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR