1,5,5-Trimethylhydantoin 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24188-14
B24188-06
B24188-06
B24188-14
B24188-22
B24188-22
AAB24188-06EA
54.99
USD
AAB24188-06
CAAAB24188-06
AAB24188-14
AAB24188-22
CAAAB24188-14
CAAAB24188-22
1,5,5-Trimethylhydantoin 98%
1,5,5-Trimethylhydantoin
Formula:
C₆H₁₀N₂O₂ Melting Pt: 161…164 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040439 CAS Number: 6851-81-6 EINECS: 229-945-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...