Quinazoline 98%

Supplier: Alfa Aesar

B24094-06 B24094-03 B24094-03 B24094-06
AAB24094-06EA 141.34 USD
AAB24094-06 CAAAB24094-03 AAB24094-03 CAAAB24094-06
Quinazoline 98%
Quinazoline

Anti-malarial agent

Formula: C₈H₆N₂
MW: 130.15 g/mol
Boiling Pt: 243 °C
Melting Pt: 45…48 °C
Flash Pt: 106 °C (224 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006712
CAS Number: 253-82-7
EINECS: 205-965-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR