N,N-Diethylacetoacetamide 97%
Supplier: Thermo Scientific Chemicals
B24019-30
B24019-18
B24019-30
B24019-18
AAB24019-18EA
31.69
USD
AAB24019-18
AAB24019-30
CAAAB24019-18
CAAAB24019-30
N,N-Diethylacetoacetamide 97%
N,N-Diethylacetoacetamide
Formula:
C₈H₁₅NO₂ MW: 157.20 g/mol Boiling Pt: 213…218 °C Melting Pt: < –73 °C Density: 0.990 Flash Pt: 94 °C(201 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00026728 CAS Number: 2235-46-3 EINECS: 218-792-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...