(2S,3S)-(+)-2,3-O-Isopropylidene-L-threitol 98%

Synonyms: (+)-2,3-O-Isopropylidene-L-threitol

B23090-06 B23090-03 B23090-06 B23090-03
AAB23090-06EA 278.82 USD
CAAAB23090-03 AAB23090-06 AAB23090-03 CAAAB23090-06
(2S,3S)-(+)-2,3-O-Isopropylidene-L-threitol 98%
(2S,3S)-(+)-2,3-O-Isopropylidene-L-threitol
Formula: C₇H₁₄O₄
Boiling Pt: 92…94 °C/0.1 mm
Melting Pt: 45…49 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00063761
CAS Number: 50622-09-8
EINECS: 256-658-6

Order Now

Specification Test Results

Optical rotation +3° (c=5 in ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR