4-Methyl-1-tetralone 97%

Synonyms: 4-Methyl-3,4-dihydronaphthalen-1(2H)-one

B21967-14 B21967-06 B21967-14 B21967-03 B21967-06 B21967-03
AAB21967-06EA 94.71 USD
AAB21967-06 CAAAB21967-06 AAB21967-14 AAB21967-03 CAAAB21967-14 CAAAB21967-03
4-Methyl-1-tetralone 97%
4-Methyl-1-tetralone
Formula: C₁₁H₁₂O
MW: 160.22 g/mol
Boiling Pt: 95 °C/1 mm
Density: 1.078
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001691
CAS Number: 19832-98-5
EINECS: 243-355-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR