Digoxin 96%

Supplier: Thermo Scientific Chemicals
Danger

B21902-06 B21902-03 B21902-03 B21902-06
AAB21902-03EA 145.52 USD
AAB21902-03 CAAAB21902-03 CAAAB21902-06 AAB21902-06
Digoxin 96%
Digoxin

A cardiac glucoside and substrate for Pgp; upregulates Pgp expression.

Formula: C₄₁H₆₄O₁₄
Melting Pt: ca 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003674
CAS Number: 20830-75-5
EINECS: 244-068-1
UN: 3462
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR