S-Propyl thioacetate 98+%

Warning

Synonyms: S-Propyl ethanethioate

B21875-14 B21875-06 B21875-14 B21875-22 B21875-22 B21875-06
AAB21875-06EA 167.07 USD
CAAAB21875-14 CAAAB21875-22 AAB21875-06 AAB21875-14 CAAAB21875-06 AAB21875-22
S-Propyl thioacetate 98+%
S-Propyl thioacetate
Formula: C₅H₁₀OS
MW: 118.2 g/mol
Boiling Pt: 137…139 °C
Density: 0.971
Storage Temperature: Ambient
MDL Number: MFCD00039937
CAS Number: 2307-10-0
EINECS: 218-984-7
UN: 3272
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR