1,2-Dibromo-1-iodo-1,2,2-trifluoroethane 97%

Warning

Synonyms: 1,2-Dibromo-1-iodotrifluoroethane

B21532-03 B21532-03 B21532-06 B21532-06
AAB21532-03EA 88.75 USD
AAB21532-03 CAAAB21532-06 CAAAB21532-03 AAB21532-06
1,2-Dibromo-1-iodo-1,2,2-trifluoroethane 97%
1,2-Dibromo-1-iodo-1,2,2-trifluoroethane
Formula: C₂Br₂F₃I
MW: 367.73 g/mol
Boiling Pt: 63…65 °C/32 mm
Density: 1.515
Storage Temperature: Ambient
MDL Number: MFCD00042521
CAS Number: 216394-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR