N-Carbamoylmaleamic acid 98%

Warning

B21487-09 B21487-18 B21487-18 B21487-09
AAB21487-18EA 122.16 USD
AAB21487-18 AAB21487-09 CAAAB21487-18 CAAAB21487-09
N-Carbamoylmaleamic acid 98%
N-Carbamoylmaleamic acid
Formula: C₅H₆N₂O₄
Melting Pt: 156…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00014790
CAS Number: 105-61-3
EINECS: 203-314-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR