5-Fluoroorotic acid monohydrate (5-FOA) 98%

Warning

Synonyms: 5-FOA5-Fluoro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate5-Fluorouracil-6-carboxylic acid monohydrate

B21290-MF B21290-03 B21290-03 B21290-MF
AAAB21290-MFEA 42.74 USD
AAAB21290-MF CAAAAB21290-MF AAAB21290-03 CAAAAB21290-03
5-Fluoroorotic acid monohydrate (5-FOA) 98%
5-Fluoroorotic acid monohydrate
Formula: C₅H₃FN₂O₄·1H₂O
MW: 174.0 g/mol (anhy)
Melting Pt: ca 255 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00150658
CAS Number: 220141-70-8
EINECS: 211-876-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR