DL-α-Phenylglycine 98%

Synonyms: H-DL-Phg-OHDL-2-PhenylglycineDl-A-Aminophenylacetic Acid

B21129-22 B21129-36 B21129-36 B21129-14 B21129-22
AAB21129-22EA 300.99 USD
AAB21129-22 AAB21129-36 CAAAB21129-36 CAAAAB21129-14 CAAAB21129-22
DL-α-Phenylglycine 98%
DL-α-Phenylglycine
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Melting Pt: ca 290 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00064402
CAS Number: 2835-06-5
EINECS: 220-608-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR