5-Methyl-5-nonanol 99%

B20810-06 B20810-14 B20810-14 B20810-06
AAB20810-14EA 303.03 USD
AAB20810-14 CAAAB20810-06 CAAAB20810-14 AAB20810-06
5-Methyl-5-nonanol 99%
5-Methyl-5-nonanol
Formula: C₁₀H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD00021844
CAS Number: 33933-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR