Cyclohexyl butyrate 99%

Warning

B20695-30 B20695-30 B20695-18 B20695-18
AAB20695-18EA 36.33 USD
AAB20695-18 CAAAB20695-30 AAB20695-30 CAAAB20695-18
Cyclohexyl butyrate 99%
Cyclohexyl butyrate
Formula: C₁₀H₁₈O₂
Boiling Pt: 90 °C/15 mm
Density: 0.940
Storage Temperature: Ambient
MDL Number: MFCD00046354
CAS Number: 1551-44-6
EINECS: 216-290-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR