3-Ethyl-2-heptanol erythro + threo 98%

Warning

B20652-03 B20652-14 B20652-06 B20652-06 B20652-14 B20652-03
AAB20652-03EA 261.39 USD
CAAAB20652-14 CAAAB20652-03 AAB20652-03 AAB20652-14 AAB20652-06 CAAAB20652-06
3-Ethyl-2-heptanol erythro + threo 98%
3-Ethyl-2-heptanol
Formula: C₉H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD00046644
CAS Number: 19780-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR