4-Decyne 98%

Warning

B20597-14 B20597-14 B20597-06 B20597-06
AAB20597-14EA 243.09 USD
AAB20597-14 CAAAB20597-06 CAAAB20597-14 AAB20597-06
4-Decyne 98%
4-Decyne
Formula: C₁₀H₁₈
Boiling Pt: 75…76 °C/20 mm
Density: 0.768
Flash Pt: 52 °C(125 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039978
CAS Number: 2384-86-3
UN: 3295
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR