(±)-4,7-Dimethyl-4-octanol 99%
|
Warning
|
B20225-14
B20225-03
B20225-03
B20225-06
B20225-14
B20225-06
AAB20225-06EA
85.56
USD
CAAAB20225-06
AAB20225-06
AAB20225-03
AAB20225-14
CAAAB20225-14
CAAAB20225-03
(±)-4,7-Dimethyl-4-octanol 99%
(±)-4,7-Dimethyl-4-octanol
Formula:
C₁₀H₂₂O Storage Temperature: Ambient |
MDL Number:
MFCD00046603 CAS Number: 19781-13-6 EINECS: 243-306-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...