Periodic acid 34% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Orthoperiodic acidortho-Periodic acid

B20172-0E B20172-36
AAB20172-36EA 139.99 USD
AAB20172-36 AAB20172-0E
Periodic acid 34% (w/w) in aqueous solution
Periodic acid
Formula: H₅IO₆
MW: 227.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011344
CAS Number: 10450-60-9
EINECS: 233-937-0
UN: 3098
ADR: "5.1",III
https://us.vwr.com/stibo/web/std.lang.all/90/77/779077.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR