1-(Hydroxymethyl)-5,5-dimethylhydantoin 97%

Warning

Synonyms: 5,5-Dimethyl-1-(hydroxymethyl)hydantoin

A19868-14 A19868-14 A19868-22 A19868-22
AAA19868-14EA 72.79 USD
CAAAA19868-22 AAA19868-14 AAA19868-22 CAAAA19868-14
1-(Hydroxymethyl)-5,5-dimethylhydantoin 97%
1-(Hydroxymethyl)-5,5-dimethylhydantoin
Formula: C₆H₁₀N₂O₃
MW: 158.16
Melting Pt: 104…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00020866
CAS Number: 116-25-6
EINECS: 204-132-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR