Bis(methylthio)methane 99%

Supplier: Thermo Scientific Chemicals

Warning

A19706-14 A19706-14
AAA19706-14EA 86.9 USD
AAA19706-14 CAAAA19706-14
Bis(methylthio)methane 99%
Bis(methylthio)methane
Formula: C₃H₈S₂
Boiling Pt: 146…147 °C
Density: 1.059
Flash Pt: 43 °C(109 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008564
CAS Number: 1618-26-4
EINECS: 216-577-9
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR