Soda lime indicator
Supplier: Thermo Scientific Chemicals
|
Danger
|
A19352-0E
A19352-36
AAA19352-36EA
59.33
USD
AAA19352-36
AAA19352-0E
Soda lime indicator
Soda lime
Storage Temperature:
Ambient |
MDL Number:
MFCD00134094 CAS Number: 8006-28-8 ADR: 8,III Merck Index: 13,08640 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...