Thulium (III) sulfate octahydrate 99%

Warning

A17296-03
CAAAAA17296-03
Thulium (III) sulfate octahydrate 99%
Thulium (III) sulphate octahydrate
Formula: O₁₂S₃Tm₂·8H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149881
CAS Number: 13778-40-0
EINECS: 243-996-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR