2-Amino-5-(trifluoromethyl)-1,3,4-thiadiazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-5-trifluoromethyl-1,3,4-thiadiazole

A17061-03 A17061-03 A17061-14 A17061-06 A17061-06 A17061-14
AAAA17061-14EA 302.99 USD
AAAA17061-14 CAAAAA17061-14 CAAAA17061-06 CAAAA17061-03 AAA17061-03 AAA17061-06
2-Amino-5-(trifluoromethyl)-1,3,4-thiadiazole 98%
2-Amino-5-(trifluoromethyl)-1,3,4-thiadiazole
Formula: C₃H₂F₃N₃S
MW: 169.13 g/mol
Melting Pt: 222…227 °C
Storage Temperature: Ambient
MDL Number: MFCD00003109
CAS Number: 10444-89-0
EINECS: 233-930-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR