2,2,6,6-Tetramethyl-4-piperidone monohydrate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,2,6,6-Tetramethyl-4-piperidone Hydrate

A16211-18 A16211-30 A16211-09 A16211-18 A16211-30 A16211-09
AAA16211-30EA 466.59 USD
CAAAA16211-30 AAA16211-30 AAA16211-18 AAA16211-09 CAAAA16211-18 CAAAA16211-09
2,2,6,6-Tetramethyl-4-piperidone monohydrate 99%
2,2,6,6-Tetramethyl-4-piperidone monohydrate
Formula: C₉H₁₇NO·1H₂O
MW: 173.26 g/mol
Melting Pt: 63…61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149393
CAS Number: 10581-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR