Ethyl vinyl ether 99% stabilized

Supplier: Thermo Scientific Chemicals
Danger

A15691-0F A15691-0F A15691-AK A15691-AK A15691-AP A15691-AP
AAA15691-0FEA 154.99 USD
AAA15691-0F CAAAA15691-AP AAA15691-AP AAAA15691-AK CAAAA15691-0F CAAAAA15691-AK
Ethyl vinyl ether 99% stabilized
Ethyl vinyl ether
Formula: C₄H₈O
Boiling Pt: 33…34 °C
Melting Pt: –115 °C
Density: 0.754
Flash Pt: -45 °C(-49 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00009248
CAS Number: 109-92-2
EINECS: 203-718-4
UN: 1302
ADR: "3",I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR