Cinnamonitrile cis- and trans- mixture 97%

Warning

Synonyms: Cinnamonitrile3-Phenylacrylonitrile

A15301-22 A15301-14 A15301-36 A15301-22 A15301-36 A15301-14
AAA15301-22EA 186.43 USD
CAAAA15301-14 CAAAA15301-36 CAAAA15301-22 AAA15301-22 AAA15301-36 AAA15301-14
Cinnamonitrile cis- and trans- mixture 97%
Cinnamonitrile
Formula: C₉H₇N
MW: 129.16 g/mol
Boiling Pt: 253…254 °C
Density: 1,029
Flash Pt: 126 °C(258 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001930
CAS Number: 4360-47-8
EINECS: 224-441-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR