Iodoacetamide 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Iodoacetamide
Stabilization:
Moistened with water ∼5-8%
Alkylating agent used in peptide mapping because it covalently binds with the thiols in cysteine to prevent disulfide bond formation
2-Iodoacetamide is used as an electrophile for covalent modification of nucleophilic residues on proteins such as cysteine, methionine and histidine. It is used to bind with thiol group of cysteine, thereby it protects the formation disulfide bonds. It is involved as an inhibitor of deubiquitinase enzymes (DUBs). Further, it acts as an alkylating sulfhydryl reagent.
Formula:
ICH₂CONH₂ MW: 184.96 g/mol Melting Pt: 92…95 °C (dried) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008028 CAS Number: 144-48-9 EINECS: 205-630-1 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...