Aluminium chloride hexahydrate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Aluminum chloride hexahydrate

A14437-0I A14437-0B A14437-30
AAA14437-0BEA 164.7 USD
AAA14437-0B AAA14437-0I AAA14437-30
Aluminium chloride hexahydrate 99%
Aluminium chloride hexahydrate
Formula: AlCl₃·6H₂O
MW: 241.43 g/mol
Melting Pt: 100 °C (dec.)
Density: 2.398
Storage Temperature: Ambient
MDL Number: MFCD00149134
CAS Number: 7784-13-6
EINECS: 231-208-1
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR