Ethoxymethylenemalononitrile 97+%

Supplier: Thermo Scientific Chemicals
Danger

A14412-36 A14412-36 A14412-14 A14412-14 A14412-22 A14412-22
AAA14412-36EA 471.74 USD
CAAAA14412-36 CAAAA14412-14 AAA14412-36 AAA14412-14 AAA14412-22 CAAAA14412-22
Ethoxymethylenemalononitrile 97+%
Ethoxymethylenemalononitrile
Formula: C₆H₆N₂O
MW: 122.13 g/mol
Boiling Pt: 160 °C/12 mm
Melting Pt: 62…68 °C
Flash Pt: 155 °C(311 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001854
CAS Number: 123-06-8
EINECS: 204-597-0
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR