(S)-(-)-4-Isopropyl-2-oxazolidinone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (4S)-2-Oxo-4-(prop-2-yl)-1,3-oxazolidine(4S)-(-)-4-Isopropyl-2-oxazolidinone(S)-4-Isopropyl-2-oxazolidinone(4S)-(-)-Isopropyl-2-oxazolidinone(4S)-4-(Prop-2-yl)-1,3-oxazolidin-2-one

A14029-03 A14029-03 A14029-06 A14029-06
AAA14029-03EA 59.29 USD
CAAAA14029-03 AAA14029-03 AAA14029-06 CAAAA14029-06
(S)-(-)-4-Isopropyl-2-oxazolidinone 98%
(S)-(-)-4-Isopropyl-2-oxazolidinone
Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00010847
CAS Number: 17016-83-0

Order Now

Specification Test Results

Optical rotation -17° (c=6 in ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR