N,N'-Diacetylhydrazine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N'-Acetylacetohydrazide
A13393-18
A13393-09
A13393-09
A13393-30
A13393-18
A13393-30
AAAA13393-30EA
682.99
USD
CAAAAA13393-30
AAAA13393-30
CAAAA13393-09
AAA13393-18
CAAAA13393-18
AAA13393-09
N,N'-Diacetylhydrazine 98%
N,N'-Diacetylhydrazine
Formula:
C₄H₈N₂O₂ MW: 116.12 g/mol Boiling Pt: 209 °C/15 mm Melting Pt: 138…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008673 CAS Number: 3148-73-0 EINECS: 221-576-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...